Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DC-SIGN/CD209 Antibody (102F10.04), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. DDX0205A488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDX0205A488 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDX0205A488 Supplier Novus Biologicals Supplier No. DDX0205A488100
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DC-SIGN/CD209 Monoclonal specifically detects DC-SIGN/CD209 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen DC-SIGN/CD209
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 102F10.04
Concentration 0.5mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence
Gene Accession No. NP_066978
Gene Alias CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1
Gene Symbols CD209
Host Species Mouse
Immunogen DC-SIGN/CD209 was used as immunogen.
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 30835
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.