Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DC-SIGN/CD209 Antibody (MM0239-4G10), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP212150G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212150G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212150G Supplier Novus Biologicals Supplier No. NBP212150G

Mouse Monoclonal Antibody

DC-SIGN/CD209 Monoclonal specifically detects DC-SIGN/CD209 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen DC-SIGN/CD209
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MM0239-4G10
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Accession No. Q9NNX6
Gene Alias CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1
Gene Symbols CD209
Host Species Mouse
Immunogen Human recombinant DC-SIGN/CD209
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 30835
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.