Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DC-SIGN/CD209 Antibody (UW60.1), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N221791APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N221791APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N221791APC7 Supplier Novus Biologicals Supplier No. NBP221791APCCY7

Mouse Monoclonal Antibody

DC-SIGN/CD209 Monoclonal specifically detects DC-SIGN/CD209 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen DC-SIGN/CD209
Applications Flow Cytometry
Classification Monoclonal
Clone UW60.1
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1
Gene Symbols CD209
Host Species Mouse
Immunogen CD209-His-tagged fusion protein.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 30835.0
Test Specificity The mouse monoclonal antibody UW60.1 recognizes human CD209 (DC-SIGN), a 44 kDa transmembrane receptor expressed on the surface of dendritic cells and macrophages.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.