Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DC-SIGN+DC-SIGNR Antibody (DC28), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB297064 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297064 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB297064 Supplier Novus Biologicals Supplier No. FAB16211A
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DC-SIGN+DC-SIGNR Monoclonal antibody specifically detects DC-SIGN+DC-SIGNR in Human samples. It is validated for Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF

Specifications

Antigen DC-SIGN+DC-SIGNR
Applications Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF
Classification Monoclonal
Clone DC28
Conjugate APC
Dilution Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF-ready
Formulation PBS
Gene Alias CD209,CD209 antigen,CDSIGN,CLEC4L,DC-SIGN,DC-SIGN1,hDC-SIGN
Host Species Mouse
Immunogen E. coli-derived recombinant human DC-SIGN, Extracellular domain
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30835
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.