Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DC-SIGNR/CD299/CLEC4M Antibody (120604), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB175258 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175258 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175258 Supplier Novus Biologicals Supplier No. FAB162K
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

DC-SIGNR/CD299/CLEC4M Monoclonal antibody specifically detects DC-SIGNR/CD299/CLEC4M in Human samples. It is validated for Flow Cytometry

Specifications

Antigen DC-SIGNR/CD299/CLEC4M
Applications Flow Cytometry
Classification Monoclonal
Clone 120604
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias CD209L1, CD209Ldendritic cell-specific ICAM-3-grabbing nonintegrin 2, CD299, CD299 antigenMGC129964, C-type lectin domain family 4, member M, DC-SIGN2CD209 antigen-like protein 1, DC-SIGNRDC-SIGN-related protein, DCSIGNRMGC47866, Dendritic cell-specific ICAM-3-grabbing non-integrin 2, HP10347, liver/lymph node-specific ICAM-3 grabbing non-integrin, Liver/lymph node-specific ICAM-3-grabbing non-integrin, L-SIGN, LSIGNC-type lectin domain family 4 member M, mannose binding C-type lectin DC-SIGNR
Host Species Mouse
Immunogen NIH-3T3 mouse embryonic fibroblast cell line transfected with human DC-SIGNR/CD299, Accession # Q9H2X3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Signal Transduction, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 10332
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.