Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DC-SIGNR/CD299/CLEC4M Antibody (16E7) - Chimeric - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP25264902 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25264902 0.2 mg
NBP25264925 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25264902 Supplier Novus Biologicals Supplier No. NBP2526490.2MG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

DC-SIGNR/CD299/CLEC4M Monoclonal specifically detects DC-SIGNR/CD299/CLEC4M in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.

Specifications

Antigen DC-SIGNR/CD299/CLEC4M
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 160000000
Conjugate Unconjugated
Dilution Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, CyTOF-ready
Formulation PBS with 0.02% Proclin 300. with 0.02% Proclin 300
Gene Alias CD209L1, CD209Ldendritic cell-specific ICAM-3-grabbing nonintegrin 2, CD299, CD299 antigenMGC129964, C-type lectin domain family 4, member M, DC-SIGN2CD209 antigen-like protein 1, DC-SIGNRDC-SIGN-related protein, DCSIGNRMGC47866, Dendritic cell-specific ICAM-3-grabbing non-integrin 2, HP10347, liver/lymph node-specific ICAM-3 grabbing non-integrin, Liver/lymph node-specific ICAM-3-grabbing non-integrin, L-SIGN, LSIGNC-type lectin domain family 4 member M, mannose binding C-type lectin DC-SIGNR
Gene Symbols CLEC4M
Host Species Rabbit
Immunogen CD299
Purification Method Protein A or G purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10332
Test Specificity Human CD299, which is an oligomeric type II transmembrane protein with a C-type lectin extracellular domain, the expression of which is restricted to immature DC, macrophages in the lung, and endothelial cells in the liver. It binds ICAM-3 and ICAM-7 to mediate the interaction of DC with T lymphocytes and endothelial cells in the initial stages of immune response and in the migratory behavior of DC. CD299 also binds the gp120 protein of HIV and the E2 envelope protein of HCV, thereby playing a role in viral infection.
Target Species Human
Content And Storage Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.