Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DC-STAMP/TM7SF4 Antibody (788524), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB373975 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB373975 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB373975 Supplier Novus Biologicals Supplier No. FAB7824Y
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DC-STAMP/TM7SF4 Monoclonal antibody specifically detects DC-STAMP/TM7SF4 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen DC-STAMP/TM7SF4
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 788524
Conjugate DyLight 680
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias dendritic cell-specific transmembrane protein, transmembrane 7 superfamily member 4
Host Species Mouse
Immunogen E. coli-derived recombinant human DC-STAMP, Asp314-Thr376, Accession # Q9H295
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81501
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.