Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCAF4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15286220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15286220 20 μL
NBP152862 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15286220 Supplier Novus Biologicals Supplier No. NBP15286220UL

Rabbit Polyclonal Antibody

DCAF4 Polyclonal specifically detects DCAF4 in Human samples. It is validated for Western Blot.

Specifications

Antigen DCAF4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q8IV10
Gene Alias DDB1 and CUL4 associated factor 4, DDB1- and CUL4-associated factor 4, DKFZp434K114, MGC20547, MGC46524, WD repeat domain 21, WD repeat domain 21A, WD repeat-containing protein 21A, WDR21, WDR21A
Gene Symbols DCAF4
Host Species Rabbit
Immunogen Synthetic peptides corresponding to WDR21A(WD repeat domain 21A) The peptide sequence was selected from the middle region of WDR21A. Peptide sequence GHRQSFGTNSDVLAQQFALMAPLLFNGCRSGEIFAIDLRCGNQGKGWKAT.
Molecular Weight of Antigen 43 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26094
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.