Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCAF4L2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP238107 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP238107 0.1 mL
NB396965 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP238107 Supplier Novus Biologicals Supplier No. NBP238107
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DCAF4L2 Polyclonal specifically detects DCAF4L2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen DCAF4L2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q8NA75
Gene Alias DDB1 and CUL4 associated factor 4-like 2, DDB1- and CUL4-associated factor 4-like protein 2, FLJ35775, MGC133227, WD repeat domain 21C, WD repeat-containing protein 21C, WDR21C
Gene Symbols DCAF4L2
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: AGGSKYGIITMRGLTTPELRVYPHK
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 138009
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.