Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCAMKL2 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB356512 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB356512 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB356512 Supplier Novus Biologicals Supplier No. NBP177128MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DCAMKL2 Polyclonal antibody specifically detects DCAMKL2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen DCAMKL2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CL2EC 2.7.11.1, CLICK-II, CLIK2, DCDC3, DCDC3BCaMK-like CREB regulatory kinase 2, DCK2DCAMKL2, DKFZp761I032, doublecortin and CaM kinase-like 2, Doublecortin domain-containing protein 3B, Doublecortin-like and CAM kinase-like 2, doublecortin-like kinase 2CLICK2, EC 2.7.11, MGC45428, serine/threonine-protein kinase DCLK2
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human DCLK2. The immunogen is located within the last 50 amino acids of DCLK2. Amino Acid Squence: PGGERAGTWRRHRD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 166614
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.