Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCAMKL2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP185282 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP185282 0.1 mL
NB406681 25ul
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP185282 Supplier Novus Biologicals Supplier No. NBP185282
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DCAMKL2 Polyclonal specifically detects DCAMKL2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen DCAMKL2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias CL2EC 2.7.11.1, CLICK-II, CLIK2, DCDC3, DCDC3BCaMK-like CREB regulatory kinase 2, DCK2DCAMKL2, DKFZp761I032, doublecortin and CaM kinase-like 2, Doublecortin domain-containing protein 3B, Doublecortin-like and CAM kinase-like 2, doublecortin-like kinase 2CLICK2, EC 2.7.11, MGC45428, serine/threonine-protein kinase DCLK2
Gene Symbols DCLK2
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:ENNMQAEVTGKLKQHFNNALPKQNSTTTGVSVIMNTALDKEGQIFCSKHCQDSGRPGMEPISPVPPSVEEIPVPGEAVPAPTPPESPTPHCPPAAPGGERAG
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 166614
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.