Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCAR/CLEC4B Antibody (349214), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB364332 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB364332 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB364332 Supplier Novus Biologicals Supplier No. FAB2757Z
Add to Cart
Add to Cart

Rat Monoclonal Antibody

DCAR/CLEC4B Monoclonal antibody specifically detects DCAR/CLEC4B in Mouse samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen DCAR/CLEC4B
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 349214
Conjugate DyLight 755
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CLECSF3, LSLCL, P47, SCGF
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse DCAR/CLEC4B, Gln42-Leu209, Accession # AAR13070
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6320
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.