Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCBLD2/ESDN Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10093567 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10093567 0.1 mg
NB017920 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10093567 Supplier Novus Biologicals Supplier No. NB10093567
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DCBLD2/ESDN Polyclonal specifically detects DCBLD2/ESDN in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen DCBLD2/ESDN
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS with 0.02% Sodium Azide
Gene Alias CLCP11700055P21Rik, CUB, LCCL and coagulation factor V/VIII-homology domains protein 1, discoidin, CUB and LCCL domain containing 2, discoidin, CUB and LCCL domain-containing protein 2, Endothelial and smooth muscle cell-derived neuropilin-like protein, ESDNcoagulation factor V/VIII-homology domains protein 1
Gene Symbols DCBLD2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues 402-419 [QDKIFQGNKDYHKDVRNN] of the human ESDN protein.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 131566
Test Specificity ESDN
Target Species Human, Mouse, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.