Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCBLD2/ESDN Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $426.00
Specifications
Antigen | DCBLD2/ESDN |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017920
![]() |
Novus Biologicals
NB10093567SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10093567
![]() |
Novus Biologicals
NB10093567 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
DCBLD2/ESDN Polyclonal specifically detects DCBLD2/ESDN in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
DCBLD2/ESDN | |
Western Blot 1:1000 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
PBS with 0.02% Sodium Azide | |
131566 | |
Synthetic peptide corresponding to residues 402-419 [QDKIFQGNKDYHKDVRNN] of the human ESDN protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Mouse, Rat, Human | |
CLCP11700055P21Rik, CUB, LCCL and coagulation factor V/VIII-homology domains protein 1, discoidin, CUB and LCCL domain containing 2, discoidin, CUB and LCCL domain-containing protein 2, Endothelial and smooth muscle cell-derived neuropilin-like protein, ESDNcoagulation factor V/VIII-homology domains protein 1 | |
DCBLD2 | |
IgG | |
Affinity Purified | |
ESDN |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title