Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCBLD2/ESDN Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB329498 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB329498 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB329498 Supplier Novus Biologicals Supplier No. NBP300160PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DCBLD2/ESDN Polyclonal antibody specifically detects DCBLD2/ESDN in Human samples. It is validated for ELISA

Specifications

Antigen DCBLD2/ESDN
Applications ELISA
Classification Polyclonal
Conjugate PE
Dilution ELISA
Formulation PBS
Gene Alias CLCP11700055P21Rik, CUB, LCCL and coagulation factor V/VIII-homology domains protein 1, discoidin, CUB and LCCL domain containing 2, discoidin, CUB and LCCL domain-containing protein 2, Endothelial and smooth muscle cell-derived neuropilin-like protein, ESDNcoagulation factor V/VIII-homology domains protein 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human DCBLD2/ESDN (Accession#: NP_563615.3; Met1-Ala482)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 131566
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.