Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCIR/CLEC4A Antibody (216110) [Janelia Fluor™ 549], Novus Biologicals™
SDP

Catalog No. NB398996 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398996 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398996 Supplier Novus Biologicals Supplier No. FAB1748I
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DCIR/CLEC4A Monoclonal antibody specifically detects DCIR/CLEC4A in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen DCIR/CLEC4A
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 216110
Conjugate Janelia Fluor 549
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CLECSF6C-type lectin superfamily member 6, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 6, C-type lectin domain family 4 member A, C-type lectin domain family 4, member A, DCIRLLIR, DDB27C-type lectin DDB27, Dendritic cell immunoreceptor, Lectin-like immunoreceptor
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human DCIR isoform 1, Gln70-Leu237, Accession # Q9UMR7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 50856
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.