Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCIR/CLEC4A Antibody (MM0237-5P20), PerCP, Novus Biologicals™
SDP

Catalog No. NBP212257P Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212257P 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212257P Supplier Novus Biologicals Supplier No. NBP212257PCP

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

DCIR/CLEC4A Monoclonal specifically detects DCIR/CLEC4A in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen DCIR/CLEC4A
Applications Flow Cytometry
Classification Monoclonal
Clone MM0237-5P20
Conjugate PerCP
Dilution Flow Cytometry
Gene Accession No. Q9UMR7
Gene Alias CLECSF6C-type lectin superfamily member 6, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 6, C-type lectin domain family 4 member A, C-type lectin domain family 4, member A, DCIRLLIR, DDB27C-type lectin DDB27, Dendritic cell immunoreceptor, Lectin-like immunoreceptor
Gene Symbols CLEC4A
Host Species Mouse
Immunogen Human recombinant DCIR/CLEC4A (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 50856
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.