Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCK Antibody (OTI3F5), PE, Novus Biologicals™
SDP

Catalog No. NBP270555PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270555PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270555PE Supplier Novus Biologicals Supplier No. NBP270555PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DCK Monoclonal specifically detects DCK in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry.

Specifications

Antigen DCK
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3F5
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias dCK, deoxycytidine kinase, EC 2.7.1, EC 2.7.1.74, MGC117410, MGC138632
Gene Symbols DCK
Host Species Mouse
Immunogen Full length recombinant protein of human DCK (NP_000779) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 1633.0
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.