Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCNP1 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB192141 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB192141 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB192141 Supplier Novus Biologicals Supplier No. NBP281911H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DCNP1 Polyclonal antibody specifically detects DCNP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen DCNP1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias C5orf20, chromosome 5 open reading frame 20, DCNP1 dendritic cell nuclear protein 1
Host Species Rabbit
Immunogen DCNP1 antibody was raised against an 18 amino acid peptide near the amino terminus of human DCNP1. The immunogen is located within the first 50 amino acids of DCNP1. Amino Acid Squence: VPGHQRLERGAGGETPEF
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 140947
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.