Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dcp1a Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB362665 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362665 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362665 Supplier Novus Biologicals Supplier No. NBP204039R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Dcp1a Polyclonal antibody specifically detects Dcp1a in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen Dcp1a
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias DCP1 decapping enzyme homolog A (S. cerevisiae), decapping enzyme hDcp1a, EC 3.-, FLJ21691, mRNA-decapping enzyme 1A, Nbla00360, putative protein product of Nbla00360, Smad4-interacting transcriptional co-activator, SMAD4IP1, SMIFHSA275986, Transcription factor SMIF
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 475 and 525 of human Dcp1a Antibody using the numbering given in entry NP_060873.4 (GeneID 55802).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55802
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.