Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCP2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017121 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017121 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017121 Supplier Novus Biologicals Supplier No. NBP281810
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DCP2 Polyclonal specifically detects DCP2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen DCP2
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 20 μg/mL
Formulation PBS
Gene Alias DCP2 decapping enzyme homolog (S. cerevisiae), EC 3.-, FLJ33245, hDpc, mRNA-decapping enzyme 2, nudix (nucleoside diphosphate linked moiety X)-type motif 20, Nudix motif 20, NUDT20Nucleoside diphosphate-linked moiety X motif 20
Gene Symbols DCP2
Host Species Rabbit
Immunogen DCP2 antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human DCP2. The immunogen is located within amino acids 340 - 390 of DCP2. Amino Acid Squence: EKKLHPRKLQDNFETDA
Molecular Weight of Antigen 46 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 167227
Test Specificity Multiple isoforms of DCP2 are known to exist.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.