Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCP2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP157442 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP157442 100 μL
NBP15744220 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP157442 Supplier Novus Biologicals Supplier No. NBP157442
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DCP2 Polyclonal specifically detects DCP2 in Human samples. It is validated for Western Blot.

Specifications

Antigen DCP2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q8IU60
Gene Alias DCP2 decapping enzyme homolog (S. cerevisiae), EC 3.-, FLJ33245, hDpc, mRNA-decapping enzyme 2, nudix (nucleoside diphosphate linked moiety X)-type motif 20, Nudix motif 20, NUDT20Nucleoside diphosphate-linked moiety X motif 20
Gene Symbols DCP2
Host Species Rabbit
Immunogen Synthetic peptides corresponding to DCP2(DCP2 decapping enzyme homolog (S. cerevisiae)) The peptide sequence was selected from the C terminal of DCP2. Peptide sequence VEKLSRTKFRHSQQLFPDGSPGDQWVKHRQPLQQKPYNNHSEMSDLLKGK.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 167227
Test Specificity Expected identity based on immunogen sequence: Equine: 100%; Human: 100%; Pig: 100%; Canine: 92%; Guinea pig: 85%; Mouse: 78%; Rat: 78%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.