Learn More
Description
Specifications
Specifications
| Antigen | DCP2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000 |
| Formulation | PBS & 2% Sucrose. with No Preservative |
| Gene Accession No. | Q8IU60 |
| Gene Alias | DCP2 decapping enzyme homolog (S. cerevisiae), EC 3.-, FLJ33245, hDpc, mRNA-decapping enzyme 2, nudix (nucleoside diphosphate linked moiety X)-type motif 20, Nudix motif 20, NUDT20Nucleoside diphosphate-linked moiety X motif 20 |
| Gene Symbols | DCP2 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
