Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCPS Antibody (OTI4H8), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB288621 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288621 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288621 Supplier Novus Biologicals Supplier No. NBP271839APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DCPS Monoclonal antibody specifically detects DCPS in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen DCPS
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4H8
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias DCS1, DCS-1, decapping enzyme, scavenger, EC 3.-, heat shock-like protein 1, HINT5, HINT-5homolog of C. elegans 7meGMP-directed hydrolase dcs-1, Hint-related 7meGMP-directed hydrolase, Histidine triad protein member 5, HSL1, HSPC015, mRNA decapping enzyme, scavenger mRNA-decapping enzyme DcpS
Host Species Mouse
Immunogen Full length human recombinant protein of human DCPS (NP_054745) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 28960
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.