Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCPS Rabbit anti-Human, DyLight 405, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB149757 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB149757 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB149757 Supplier Novus Biologicals Supplier No. NBP297929V

Rabbit Polyclonal Antibody

DCPS Polyclonal specifically detects DCPS in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen DCPS
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DCS1, DCS-1, decapping enzyme, scavenger, EC 3.-, heat shock-like protein 1, HINT5, HINT-5homolog of C. elegans 7meGMP-directed hydrolase dcs-1, Hint-related 7meGMP-directed hydrolase, Histidine triad protein member 5, HSL1, HSPC015, mRNA decapping enzyme, scavenger mRNA-decapping enzyme DcpS
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human DCPS fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28960
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.