Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DcR3/TNFRSF6B Antibody, Novus Biologicals™
SDP

Catalog No. NBP176909SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176909SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176909SS Supplier Novus Biologicals Supplier No. NBP176909SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DcR3/TNFRSF6B Polyclonal specifically detects DcR3/TNFRSF6B in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen DcR3/TNFRSF6B
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1 ug/ml
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. O95407
Gene Alias DCR3Decoy receptor for Fas ligand, Decoy receptor 3, M68DJ583P15.1.1, TR6DcR3, tumor necrosis factor receptor superfamily member 6B, tumor necrosis factor receptor superfamily, member 6b, decoy
Gene Symbols TNFRSF6B
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to amino acids near the amino terminus of human DcR3 precursor. The immunogen is located within the first 50 amino acids of DcR3. Amino Acid Squence: AETPTYPWRDAETGER
Molecular Weight of Antigen 33 kDa
Purification Method Ion exchange chromatography
Quantity 0.025 mg
Research Discipline Apoptosis, Cell Biology, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 8771
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.