Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCUN1D1 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB041582 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB041582 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB041582 Supplier Novus Biologicals Supplier No. NBP296995C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DCUN1D1 Polyclonal specifically detects DCUN1D1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen DCUN1D1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias DCN1, defective in cullin neddylation 1, domain containing 1 (S. cerevisiae), DCN1-like protein 1, DCUN1L1DCUN1 domain-containing protein 1, Defective in cullin neddylation protein 1-like protein 1, RP42 homolog, RP42Squamous cell carcinoma-related oncogene, SCCROSCRO, Tes3
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant DCUN1D1 (NP_065691.2; Met1-Val259).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 54165
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.