Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DCUN1D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519284
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519284 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519284 Supplier Invitrogen™ Supplier No. PA519284
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine and mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

Part of an E3 ubiquitin ligase complex for neddylation. Promotes neddylation of cullin components of E3 cullin-RING ubiquitin ligase complexes. Acts by binding to cullin-RBX1 complexes in the cytoplasm and promoting their nuclear translocation, enhancing recruitment of E2-NEDD8 (UBE2M-NEDD8) thioester to the complex, and optimizing the orientation of proteins in the complex to allow efficient transfer of NEDD8 from the E2 to the cullin substrates. Involved in the release of inhibitory effets of CAND1 on cullin-RING ligase E3 complex assembly and activity (PubMed:25349211, PubMed:28581483). Acts also as an oncogene facilitating malignant transformation and carcinogenic progression. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCUN1D1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene DCUN1D1
Gene Accession No. Q96GG9
Gene Alias DCN1; DCN1, defective in cullin neddylation 1, domain containing 1; DCN1, defective in cullin neddylation 1, domain containing 1 (S. cerevisiae); DCN1-like protein 1; DCNL1; DCUN1 domain-containing protein 1; Dcun1d1; DCUN1L1; defective in cullin neddylation 1 domain containing 1; defective in cullin neddylation protein 1-like protein 1; pTes3; Rp42; RP42 homolog; SCCRO; SCRO; squamous cell carcinoma related oncogene; squamous cell carcinoma-related oncogene; Tes3; testis derived transcript 3; testis-specific protein 3
Gene Symbols DCUN1D1
Host Species Goat
Immunogen Synthetic peptide sequence (RPQIAGTKSTT) corresponding to the C-terminus amino acids of DCUN1D1 (aa 248-259).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54165
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.