Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCUN1D2 Antibody (001), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB333754 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333754 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333754 Supplier Novus Biologicals Supplier No. NBP290306APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

DCUN1D2 Monoclonal antibody specifically detects DCUN1D2 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen DCUN1D2
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 001
Conjugate APC
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation PBS
Gene Alias C13orf17, chromosome 13 open reading frame 17, DCN1, defective in cullin neddylation 1, domain containing 2 (S. cerevisiae), DCN1-like protein 2, DCUN1 domain-containing protein 2, DCUN1L2, Defective in cullin neddylation protein 1-like protein 2, FLJ10704, FLJ20092
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Human DCUN1D2 (Accession#: NP_001014305.1, Met1-Phe259)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55208
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.