Learn More
Description
Specifications
Specifications
| Antigen | DCUN1D4 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.61mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 |
| Formulation | PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal |
| Gene Accession No. | Q92564 |
| Gene Alias | DCN1, defective in cullin neddylation 1, domain containing 4 (S. cerevisiae), DCN1-like protein 4, DCUN1 domain-containing protein 4, FLJ42355, KIAA0276Defective in cullin neddylation protein 1-like protein 4 |
| Gene Symbols | DCUN1D4 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
