Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCXR Monoclonal Antibody (4G4AF5)
GREENER_CHOICE

Catalog No. A21996
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21996 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A21996 Supplier LIFE TECHNOLOGIES Supplier No. A21996
Only null left
Add to Cart
Add to Cart

DCXR Monoclonal Antibody (4G4AF5)

DCXR Monoclonal Antibody for Western Blot, ICC/IF, IP, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen DCXR
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4G4AF5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation HEPES buffered saline with 0.02% sodium azide
Gene DCXR
Gene Accession No. Q7Z4W1
Gene Alias 0610038K04Rik; 1810027P18Rik; carbonyl reductase; carbonyl reductase 2; carbonyl reductase II; DCR; Dcxr; diacetyl/L-xylulose reductase; dicarbonyl and L-xylulose reductase; dicarbonyl L-xylulose reductase; dicarbonyl/L-xylulose reductase; Glb; HCR2; HCRII; KI P34H; KID; KIDCR; Kidney dicarbonyl reductase; L-xylulose reductase; P34H; PNTSU; SDR20C1; Short chain dehydrogenase/reductase family 20C member 1; short chain dehydrogenase/reductase family 20C, member 1; sperm surface protein P34H; XR
Gene Symbols DCXR
Host Species Mouse
Immunogen Human liver mitochondria.
Purification Method purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51181
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.