Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDA1 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB190870 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB190870 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB190870 Supplier Novus Biologicals Supplier No. NBP281982H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DDA1 Polyclonal antibody specifically detects DDA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen DDA1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias C19orf58, cross-immune reaction antigen PCIA1, DET1 and DDB1 associated 1, DET1- and DDB1-associated protein 1, MGC2594, PCIA-1, PCIA1chromosome 19 open reading frame 58, Placenta cross-immune reaction antigen 1
Host Species Rabbit
Immunogen DDA1 antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human DDA1. The immunogen is located within the last 50 amino acids of DDA1. Amino Acid Squence: SAPPRKVARTDSPDMHEDT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79016
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.