Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDB1 Antibody (C.15200221), Novus Biologicals™
SDP

Catalog No. NBP262791 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP262791 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP262791 Supplier Novus Biologicals Supplier No. NBP262791
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DDB1 Monoclonal specifically detects DDB1 in Human samples. It is validated for Western Blot.

Specifications

Antigen DDB1
Applications Western Blot
Classification Monoclonal
Clone C.15200221
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS with 0.05% Sodium Azide
Gene Alias damage-specific DNA binding protein 1 (127kD), damage-specific DNA binding protein 1, 127kDa, Damage-specific DNA-binding protein 1, DDB p127 subunit, DDBa, DNA damage-binding protein 1, DNA damage-binding protein a, HBV X-associated protein 1, UV-damaged DNA-binding factor, UV-damaged DNA-binding protein 1, UV-DDB 1, UV-DDB1, XAP1, XAP-1, Xeroderma pigmentosum group E-complementing protein, XPCe, XPE, XPE-BF, XPE-binding factor
Gene Symbols DDB1
Host Species Mouse
Immunogen Human DDB1 (Damage-Specific DNA Binding Protein 1)
Purification Method Protein A purified
Quantity 50 μg
Research Discipline DNA Repair, Nucleotide Excision Repair
Primary or Secondary Primary
Gene ID (Entrez) 1642
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.