Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDB1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227207 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227207 0.1 mg
NBP227207SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227207 Supplier Novus Biologicals Supplier No. NBP227207
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DDB1 Polyclonal specifically detects DDB1 in Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Feline, Primate samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen DDB1
Applications Western Blot, Flow Cytometry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4ug/ml∼, Flow Cytometry 0.2ug/10^6cells
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q16531
Gene Alias damage-specific DNA binding protein 1 (127kD), damage-specific DNA binding protein 1, 127kDa, Damage-specific DNA-binding protein 1, DDB p127 subunit, DDBa, DNA damage-binding protein 1, DNA damage-binding protein a, HBV X-associated protein 1, UV-damaged DNA-binding factor, UV-damaged DNA-binding protein 1, UV-DDB 1, UV-DDB1, XAP1, XAP-1, Xeroderma pigmentosum group E-complementing protein, XPCe, XPE, XPE-BF, XPE-binding factor
Gene Symbols DDB1
Host Species Rabbit
Immunogen A portion of amino acids 385-435 from human DDB1 was used as the immunogen for this antibody.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline DNA Repair, Nucleotide Excision Repair
Primary or Secondary Primary
Gene ID (Entrez) 1642
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, panda, cow, dog, old world monkey, horse, african bush elephant, crab-eating macaque, rhesus monkey, mouse, chimpanzee, orangutan, rat, pig, and european pole-cat, 94% in guinea pig, Chinese hamster, platypus, and zebra finch, 89% in Carolina anole, zebrafish, mole rat, cichlid fish, and salmon, 83% in chicken, western clawed frog, and african clawed frog, 78% in wild turkey, and 61% homolgous in common grape vine, soya bean, and tomato.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Feline, Other, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.