Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDB1 Mouse anti-Human, Mouse, Rat, Clone: 4F6C6, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | DDB1 |
|---|---|
| Clone | 4F6C6 |
| Concentration | 0.43 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6453
![]()
|
Proteintech Group Inc
660101IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is specific to DDB1.
This gene encodes the large subunit of DNA damage-binding protein which is a heterodimer composed of a large and a small subunit. This protein functions in nucleotide-excision repair. Its defective activity causes the repair defect in the patients with xeroderma pigmentosum complementation group E (XPE). However, it remains for mutation analysis to demonstrate whether the defect in XPE patients is in this gene or the gene encoding the small subunit. In addition, Best vitelliform mascular dystrophy is mapped to the same region as this gene on 11q, but no sequence alternations of this gene are demonstrated in Best disease patients.Specifications
| DDB1 | |
| 0.43 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| DDB p127 subunit, DDB1, DDBA, DNA damage binding protein 1, DNA damage binding protein a, HBV X associated protein 1, UV damaged DNA binding factor, UV DDB 1, UV DDB1, XAP 1, XAP1, XPCE, XPE, XPE BF, XPE binding factor | |
| Ddb1 | |
| IgG2b | |
| Protein A | |
| Antibody |
| 4F6C6 | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| Q16531, Q3U1J4, Q9ESW0 | |
| 13194, 1642, 64470 | |
| DDB1 Fusion Protein Ag17903 | |
| Primary | |
| -20°C | |
| DDB1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title