Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDHD1 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB144526 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB144526 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB144526 Supplier Novus Biologicals Supplier No. NBP305949G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DDHD1 Polyclonal specifically detects DDHD1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen DDHD1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DDHD domain containing 1, DDHD domain-containing protein 1, EC 3.1.1.-, FLJ34209, FLJ42555, KIAA1705, PA-PLA1, Phosphatidic acid-preferring phospholipase A1 homolog, phosphatidic acid-preferring phospholipase A1-like protein, phospholipase DDHD1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human DDHD1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 80821
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.