Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDIT4L Antibody (DDIT-03), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB002676 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002676 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002676 Supplier Novus Biologicals Supplier No. NBP262218AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DDIT4L Monoclonal specifically detects DDIT4L in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen DDIT4L
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone DDIT-03
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DNA damage-inducible transcript 4-like protein, DNA-damage-inducible transcript 4-like, HIF-1 responsive protein RTP801-like, homolog of mouse SMHS1, Protein regulated in development and DNA damage response 2, REDD-2, REDD2Rtp801L, regulated in development and DNA damage response 2, RTP801L
Gene Symbols DDIT4L
Host Species Mouse
Immunogen N-terminal recombinant fragment of human DDIT4L (amino acids 2-98)
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 115265
Test Specificity The antibody MEM-18 reacts with CD14, a 53-55 kDa GPI (glycosylphosphatidylinositol)-linked membrane glycoprotein expressed on monocytes, macrophages and weakly on granulocytes; also expressed by most tissue macrophages. In human, the epitope recognized by MEM-18 is located between amino acids 57-64.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.