Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDR1 Antibody (017), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB306846 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB306846 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB306846 Supplier Novus Biologicals Supplier No. NBP289617MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

DDR1 Monoclonal antibody specifically detects DDR1 in Human samples. It is validated for ELISA

Specifications

Antigen DDR1
Applications ELISA
Classification Monoclonal
Clone 017
Conjugate mFluor Violet 450 SE
Dilution Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias CD167 antigen-like family member A, CD167a antigen, DDR, discoidin domain receptor family, member 1, discoidin domain receptor tyrosine kinase 1, Discoidin receptor tyrosine kinase, EC 2.7.10, EC 2.7.10.1, EDDR1, ENTRK4, Epithelial discoidin domain receptor 1, HGK2, MCK10, MCK-10, NTRK4, Protein-tyrosine kinase 3A, PTK3A, PTK3A protein tyrosine kinase 3A, receptor, type 4, TRK E
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human DDR1 (Accession#: NP_001945.3; Met1-Thr416).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Signaling
Primary or Secondary Primary
Gene ID (Entrez) 780
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.