Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDR2 Antibody (102), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB075840 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB075840 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB075840 Supplier Novus Biologicals Supplier No. NBP290632UV
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

DDR2 Monoclonal specifically detects DDR2 in Mouse samples. It is validated for ELISA.

Specifications

Antigen DDR2
Applications ELISA
Classification Monoclonal
Clone 102
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Gene Alias CD167 antigen-like family member B, CD167b antigen, Discoidin domain receptor 2, discoidin domain receptor family, member 2, discoidin domain receptor tyrosine kinase 2, discoidin domain-containing receptor 2, EC 2.7.10, EC 2.7.10.1, hydroxyaryl-protein kinase, migration-inducing gene 16 protein, neurotrophic tyrosine kinase receptor related 3, Neurotrophic tyrosine kinase, receptor-related 3, NTRKR3cell migration-inducing protein 20, Receptor protein-tyrosine kinase TKT, TKTMIG20a, TYRO10, Tyrosine-protein kinase TYRO10, tyrosylprotein kinase
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse DDR2 (NP_072075.2; Met1-Arg399).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4921
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.