Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDR2 Antibody (290804), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB364832 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB364832 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB364832 Supplier Novus Biologicals Supplier No. FAB25381J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DDR2 Monoclonal antibody specifically detects DDR2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-reported

Specifications

Antigen DDR2
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 290804
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, Immunohistochemistry, CyTOF-reported
Formulation 50mM Sodium Borate
Gene Alias CD167 antigen-like family member B, CD167b antigen, Discoidin domain receptor 2, discoidin domain receptor family, member 2, discoidin domain receptor tyrosine kinase 2, discoidin domain-containing receptor 2, EC 2.7.10, EC 2.7.10.1, hydroxyaryl-protein kinase, migration-inducing gene 16 protein, neurotrophic tyrosine kinase receptor related 3, Neurotrophic tyrosine kinase, receptor-related 3, NTRKR3cell migration-inducing protein 20, Receptor protein-tyrosine kinase TKT, TKTMIG20a, TYRO10, Tyrosine-protein kinase TYRO10, tyrosylprotein kinase
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human DDR2, Gln24-Arg399, Accession # Q16832
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, CD Markers, Immunology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4921
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.