Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDT Antibody (OTI1H3), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP270562R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270562R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270562R Supplier Novus Biologicals Supplier No. NBP270562R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DDT Monoclonal specifically detects DDT in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen DDT
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1H3
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias D-dopachrome decarboxylase, D-dopachrome tautomeraseDDCT, EC 4.1.1.84, Phenylpyruvate tautomerase II
Gene Symbols DDT
Host Species Mouse
Immunogen Full length recombinant protein of human DDT (NP_001346) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1652.0
Target Species Human, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.