Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX17 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB20352A532 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB20352A532 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB20352A532 Supplier Novus Biologicals Supplier No. NB200352AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DDX17 Polyclonal specifically detects DDX17 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen DDX17
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DEAD (Asp-Glu-Ala-Asp) box polypeptide 17, DEAD box protein 17, DEAD box protein p72, DEAD/H box 17, DKFZp761H2016, EC 3.6.1, EC 3.6.4.13, P72DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 17 (72kD), probable ATP-dependent RNA helicase DDX17, RH70, RNA-dependent helicase p72
Gene Symbols DDX17
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 550 and the C-terminus (residue 613) of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 17 using the numbering given in entry AAC50787.1 (GeneID 10521).
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10521
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.