Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX18 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP227353SS

 View more versions of this product

Catalog No. NBP227353SS


Only null left
Add to Cart

Description

Description

DDX18 Polyclonal specifically detects DDX18 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

DDX18
Polyclonal
Unconjugated
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
DDX18
A portion of amino acids 20-80 of human DDX18 was used as the immunogen for the antibody.
0.025 mg
Core ESC Like Genes, Stem Cell Markers
8886
Human
Purified
Western Blot, Immunohistochemistry (Paraffin)
0.5 mg/mL
Western Blot 2-4ug/ml∼, Immunohistochemistry-Paraffin 5-10ug/ml
DEAD (Asp-Glu-Ala-Asp) box polypeptide 18, DEAD box protein 18, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 18 (Myc-regulated), EC 3.6.1, EC 3.6.4.13, FLJ33908, MrDbATP-dependent RNA helicase DDX18, Myc-regulated DEAD box protein
Rabbit
Protein A purified
RUO
Primary
The amino acid sequence used as immunogen for the DDX18 antibody is 100% homologous in human and 64% homologous in mouse.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only