Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX18 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $402.00

Specifications

Antigen DDX18
Concentration 0.5 mg/mL
Dilution Western Blot 2-4ug/ml∼, Immunohistochemistry-Paraffin 5-10ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227353SS
SDP
View Documents
Novus Biologicals
NBP227353SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP227353
SDP
View Documents
Novus Biologicals
NBP227353
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

DDX18 Polyclonal specifically detects DDX18 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

DDX18
Western Blot 2-4ug/ml∼, Immunohistochemistry-Paraffin 5-10ug/ml
Polyclonal
Purified
RUO
Human
DEAD (Asp-Glu-Ala-Asp) box polypeptide 18, DEAD box protein 18, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 18 (Myc-regulated), EC 3.6.1, EC 3.6.4.13, FLJ33908, MrDbATP-dependent RNA helicase DDX18, Myc-regulated DEAD box protein
DDX18
IgG
Protein A purified
The amino acid sequence used as immunogen for the DDX18 antibody is 100% homologous in human and 64% homologous in mouse.
0.5 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Core ESC Like Genes, Stem Cell Markers
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
8886
A portion of amino acids 20-80 of human DDX18 was used as the immunogen for the antibody.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.