Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX18 Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7140323
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-8091 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-8091 Supplier Bethyl Laboratories, Inc Supplier No. A300635AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

ATP-dependent RNA helicase DDX18 belongs to theDEAD box helicase family, which contains one helicase ATP-binding domain and one helicase C-terminal domain. DDX18 is induced by Myc and Max and is involved in splicing and nuclear export. It is reported to be differentially expressed during viral latency and at early times after induction into active replication.

Specifications

Antigen DDX18
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene DDX18
Gene Accession No. Q9NVP1
Gene Alias MrDb, ATP-dependent RNA helicase DDX18, DEAD box protein 18, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 18 (Myc-regulated), Myc-regulated DEAD box protein
Gene Symbols DDX18
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8886
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.