Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX21 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB1001718SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001718SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001718SS Supplier Novus Biologicals Supplier No. NB1001718SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

DDX21 Polyclonal specifically detects DDX21 in Human, Mouse, Pig, Zebrafish samples. It is validated for Chemotaxis, ChIP Assay, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Western Blot.

Specifications

Antigen DDX21
Applications Chemotaxis, ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Chromatin Immunoprecipitation reported in scientific literature (PMID 32231306), Flow Cytometry 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunoprecipitation 2 - 10 ug/mg of lysate, Immunohistochemistry-Paraffin 1:10-1:500, Chromatin Immunoprecipitation (ChIP), Knockdown Validated reported in scientific literature (PMID 35440492)
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q9NR30
Gene Alias DEAD (Asp-Glu-Ala-Asp) box polypeptide 21, DEAD box protein 21, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 21, DKFZp686F21172, EC 3.6.1, EC 3.6.4.13, Gu protein, GUA, gu-alpha, GURDB, nucleolar RNA helicase 2, Nucleolar RNA helicase Gu, Nucleolar RNA helicase II, RH II/Gu, RH-II/GU, RH-II/GuA, RNA helicase II/Gu alpha
Gene Symbols DDX21
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 725 and the C-terminus (residue 783) of human DEAD/H (Asp-Glu-Ala-Asp/His) Box Polypeptide 21 using the numbering given in entry NP_004719.2 (GeneID 9188).
Molecular Weight of Antigen 87 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9188
Target Species Human, Mouse, Pig, Zebrafish
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.