Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX21 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB044659 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB044659 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB044659 Supplier Novus Biologicals Supplier No. NBP297537F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DDX21 Polyclonal specifically detects DDX21 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen DDX21
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias DEAD (Asp-Glu-Ala-Asp) box polypeptide 21, DEAD box protein 21, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 21, DKFZp686F21172, EC 3.6.1, EC 3.6.4.13, Gu protein, GUA, gu-alpha, GURDB, nucleolar RNA helicase 2, Nucleolar RNA helicase Gu, Nucleolar RNA helicase II, RH II/Gu, RH-II/GU, RH-II/GuA, RNA helicase II/Gu alpha
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human DDX21 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9188
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.