Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX23 Antibody, Novus Biologicals™
SDP

Catalog No. NBP216129 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP216129 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP216129 Supplier Novus Biologicals Supplier No. NBP216129
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DDX23 Polyclonal specifically detects DDX23 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen DDX23
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9BUQ8
Gene Alias 100 kDa U5 snRNP-specific protein, DEAD (Asp-Glu-Ala-Asp) box polypeptide 23, DEAD box protein 23, EC 3.6.1, EC 3.6.4.13, MGC8416, probable ATP-dependent RNA helicase DDX23, prp28, PRP28 homolog, PRP28 homolog, yeast, PRP28p homolog, PRPF28, U5 snRNP 100 kD protein, U5-100K, U5-100kD
Gene Symbols DDX23
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human DDX23. The exact sequence is proprietary.
Molecular Weight of Antigen 96 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 9416
Test Specificity Human, Mouse. Expected to cross react based on sequence identity: Rat (100%), Zebrafish (91%), Bovine (100%).
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.