Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX23 Antibody, Novus Biologicals™
SDP

Catalog No. NB1002223 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002223 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002223 Supplier Novus Biologicals Supplier No. NB1002223
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DDX23 Polyclonal specifically detects DDX23 in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen DDX23
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_004809.2
Gene Alias 100 kDa U5 snRNP-specific protein, DEAD (Asp-Glu-Ala-Asp) box polypeptide 23, DEAD box protein 23, EC 3.6.1, EC 3.6.4.13, MGC8416, probable ATP-dependent RNA helicase DDX23, prp28, PRP28 homolog, PRP28 homolog, yeast, PRP28p homolog, PRPF28, U5 snRNP 100 kD protein, U5-100K, U5-100kD
Gene Symbols DDX23
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 775 and the C-terminus (residue 820) of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 23 using the numbering given in entry NP_004809.2 (GeneID 9416).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9416
Test Specificity Human, Mouse. Based on 100% sequence identity, this antibody is predicted to react with Rat, Zebrafish, X. tropicalis, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Transparent sea squirt, West Indian ocean coelacanth, Medaka fish and Spotted green pufferfish.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.