Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDX24 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB292479 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292479 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292479 Supplier Novus Biologicals Supplier No. NB1002225F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DDX24 Polyclonal specifically detects DDX24 in Human samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen DDX24
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias ATP-dependent RNA helicase DDX24, DEAD (Asp-Glu-Ala-Asp) box polypeptide 24, DEAD box protein 24, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 24, EC 3.6.1, EC 3.6.4.13, S. cerevisiae CHL1-like helicase
Host Species Rabbit
Immunogen Maps to a region between residue 325 and 375 of human DEAD (Asp-Glu-Ala-Asp) box polypeptide 24 using the numbering given in entry NP_065147.1 (GeneID 57062).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 57062
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.